missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD126 Mouse anti-Human, PerCP-eFluor 710, Clone: 47.7G7.1F2, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $381.48

Specifications

Antigen CD126
Clone 47.7G7.1F2
Host Species Murine
Gene Alias Interleukin-6 Receptor alpha, IL-6Ra
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016009 Life Technologies
LS46126941
25 Tests Each for $157.08
5016010 Life Technologies
LS46126942
100 Tests Each for $381.48
Specifications

Description

This monoclonal antibody 47.7G7.1F2 reacts with human CD126, also known as IL-6 receptor alpha. It associates with the gp130 chain to form the high-affinity receptor for IL-6. Although gp130 is ubiquitously expressed, CD126 is only found on mature T cells, activated B cells, macrophages, hepatocytes, and myeloma cells. Upon binding to IL-6, two heterotrimeric complexes of receptor and ligand associate into a hexamer and are then internalized, initializing Jak/STAT signaling pathways. IL-6 induces the proliferation and survival of T cells, Th2 differentiation, and acute-phase protein production in the liver. A soluble form of CD126 lacking the transmembrane domain is also present and can be generated by cleavage at the cell surface or alternative splicing. Soluble CD126 can bind IL-6 and activate cells expressing only gp130 through trans-presentation.
Description & Specifications

Specifications

CD126
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
47.7G7.1F2
Interleukin-6 Receptor alpha, IL-6Ra
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test