Please login to your online account to display your discounted pricing

CD123 Rat anti-Mouse, APC, Clone: 5B11, eBioscience™

Rat Monoclonal Antibody

$187.05 - $346.55

Specifications

Antigen CD123
Clone 5B11
Host Species Rat
Gene Alias 'Interleukin-3 Receptor alpha, IL-3Ra
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015063

 
life technologies
LS17123181
50μg Each for $187.05

5015064

 
life technologies
LS17123182
100μg Each for $346.55
Description & Specifications

Specifications

Antigen CD123
Clone 5B11
Host Species Rat
Gene Alias 'Interleukin-3 Receptor alpha, IL-3Ra
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 5B11 monoclonal antibody reacts with mouse CD123, the alpha chain of the IL-3 receptor. This 60-70kDa transmembrane protein binds to IL-3 with low affinity by itself and when associated with either CD131 (common beta chain) or AIC2A (IL-3beta) binds IL-3 with high affinity. CD123 does not transduce any intracellular signals upon binding IL-3 and requires the beta chain for this function.