Please login to your online account to display your discounted pricing

CD123 Mouse anti-Human, PE-Cy5, Clone: 6H6, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $404.55

Specifications

Antigen CD123
Clone 6H6
Host Species Mouse
Gene Alias Interleukin-3 Receptor alpha, IL-3Ra
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122923

 
affymetrix
15123942
100 tests Each for $404.55

501128982

 
affymetrix
15123941
25 tests Each for $158.05
Description & Specifications

Specifications

Antigen CD123
Clone 6H6
Host Species Mouse
Gene Alias Interleukin-3 Receptor alpha, IL-3Ra
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The 6H6 monoclonal antibody reacts with human CD123, the alpha chain of the IL-3 receptor. This 60-70kDa transmembrane protein binds to IL-3 with low affinity by itself, and when associated with CD131 (common beta chain) binds IL-3 with high affinity. CD123 is expressed by myeloid precursors, macrophages, dendritic cells, mast cells, basophils, and megakaryocytes.