missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD123 Mouse anti-Human, Clone: 6H6, eBioscience™

Mouse Monoclonal Antibody

$85.55 - $143.55

Specifications

Antigen CD123
Clone 6H6
Host Species Murine
Gene Alias 'Interleukin-3 Receptor alpha, IL-3Ra
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012851 Life Technologies
LS14123980
25μg Each for $85.55
5012852 Life Technologies
LS14123982
100μg Each for $143.55
Specifications

Description

The 6H6 monoclonal antibody reacts with human CD123, the α chain of the IL-3 receptor. This 60-70kDa transmembrane protein binds to IL-3 with low affinity by itself, and when associated with CD131 (common β chain) binds IL-3 with high affinity. CD123 is expressed by myeloid precursors, macrophages, dendritic cells, mast cells, basophils, and megakaryocytes.
Description & Specifications

Specifications

CD123
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
6H6
'Interleukin-3 Receptor alpha, IL-3Ra
Flow Cytometry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen)
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL