missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD123 Mouse anti-Human, Biotin, Clone: 6H6, eBioscience™

Mouse Monoclonal Antibody

$91.08 - $209.88

Specifications

Antigen CD123
Clone 6H6
Host Species Murine
Gene Alias 'Interleukin-3 Receptor alpha, IL-3Ra
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011738 Life Technologies
LS13123980
25μg Each for $91.08
5011739 Life Technologies
LS13123982
100μg Each for $209.88
Specifications

Description

The 6H6 monoclonal antibody reacts with human CD123, the α chain of the IL-3 receptor. This 60-70kDa transmembrane protein binds to IL-3 with low affinity by itself, and when associated with CD131 (common β chain) binds IL-3 with high affinity. CD123 is expressed by myeloid precursors, macrophages, dendritic cells, mast cells, basophils, and megakaryocytes.
Description & Specifications

Specifications

CD123
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
6H6
'Interleukin-3 Receptor alpha, IL-3Ra
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL