Please login to your online account to display your discounted pricing

CD122 Rat anti-Mouse, PE, Clone: 5H4, eBioscience™

Rat Monoclonal Antibody

$230.55 - $462.55

Specifications

Antigen CD122
Clone 5H4
Host Species Rat
Gene Alias Interleukin-2 Receptor beta, IL-2Rb
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010587

 
affymetrix
12122181
50μg Each for $230.55

5010588

 
affymetrix
12122182
100μg Each for $288.55

5010589

 
affymetrix
12122183
200μg Each for $462.55
Description & Specifications

Specifications

Antigen CD122
Clone 5H4
Host Species Rat
Gene Alias Interleukin-2 Receptor beta, IL-2Rb
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 5H4 antibody reacts with mouse CD122, the 90-110kDa interleukin-2 receptor beta chain. CD122 is expressed by NK cells and some T cells in the periphery and is upregulated by activation. CD122 associates with CD25 and CD132 (common γ chain) to form the high affinity IL-2 receptor. Binding of IL-2 to CD122 is not inhibited by the 5H4 antibody.