missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD122 Rat anti-Mouse, Biotin, Clone: TM-b1 (TM-beta1), eBioscience™

Rat Monoclonal Antibody

$129.05 - $223.08

Specifications

Antigen CD122
Clone TM-b1 (TM-beta1)
Host Species Rat
Gene Alias Interleukin-2 Receptor beta, IL-2Rb
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011732 Life Technologies
LS13122281
50μg Each for $129.05
5011733 Life Technologies
LS13122282
100μg Each for $223.08
Specifications

Description

The TM-b1 monoclonal antibody reacts with mouse CD122, the 90-110kDa interleukin-2 receptor beta chain (IL-2R beta). CD122 is expressed by NK cells and some T cells in the periphery, and is upregulated by activation. CD122 associates with CD25 and CD132 (common gamma chain) to form the high affinity IL-2 receptor. High affinity binding of IL-2 to the IL-2 receptor is inhibited by the TM-b1 antibody.
Description & Specifications

Specifications

CD122
Rat
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
TM-b1 (TM-beta1)
Interleukin-2 Receptor beta, IL-2Rb
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer