missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD122 Rat anti-Mouse, Biotin, Clone: 5H4, eBioscience™

Rat Monoclonal Antibody

$117.48 - $355.08

Specifications

Antigen CD122
Clone 5H4
Host Species Rat
Gene Alias Interleukin-2 Receptor beta, IL-2Rb
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011729 Life Technologies
LS13122181
50μg Each for $117.48
5011730 Life Technologies
LS13122182
100μg Each for $223.08
5011731 Life Technologies
LS13122185
500μg Each for $355.08
Specifications

Description

The 5H4 antibody reacts with mouse CD122, the 90-110kDa interleukin-2 receptor beta chain. CD122 is expressed by NK cells and some T cells in the periphery and is upregulated by activation. CD122 associates with CD25 and CD132 (common γ chain) to form the high affinity IL-2 receptor. Binding of IL-2 to CD122 is not inhibited by the 5H4 antibody.
Description & Specifications

Specifications

CD122
Rat
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
5H4
Interleukin-2 Receptor beta, IL-2Rb
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer