Please login to your online account to display your discounted pricing

CD122 Rat anti-Mouse, APC, Clone: TM-beta1, eBioscience™

Rat Monoclonal Antibody

$158.05 - $375.55

Specifications

Antigen CD122
Clone TM-b1 (TM-beta1)
Host Species Rat
Gene Alias Interleukin-2 Receptor beta
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123071

 
life technologies
LS17122280
25μg Each for $158.05

501123072

 
life technologies
LS17122282
100μg Each for $375.55
Description & Specifications

Specifications

Antigen CD122
Clone TM-b1 (TM-beta1)
Host Species Rat
Gene Alias Interleukin-2 Receptor beta
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The TM-b1 monoclonal antibody reacts with mouse CD122, the 90-110kDa interleukin-2 receptor beta chain (IL-2R beta). CD122 is expressed by NK cells and some T cells in the periphery, and is upregulated by activation. CD122 associates with CD25 and CD132 (common gamma chain) to form the high affinity IL-2 receptor. High affinity binding of IL-2 to the IL-2 receptor is inhibited by the TM-b1 antibody.