missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD122 Mouse anti-Human, FITC, Clone: TU27, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $404.55

Specifications

Antigen CD122
Clone TU27
Host Species Murine
Gene Alias Interleukin-2 Receptor beta, IL-2Rb, IL-2Rp75
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122092 Life Technologies
LS11122841
25 Tests Each for $172.55
501122093 Life Technologies
LS11122842
100 Tests Each for $404.55
Specifications

Description

This TU27 monoclonal antibody reacts with human CD122, which is also known as interleukin-2 receptor beta (IL-2R beta). This receptor is expressed on NK cells, activated T cells, some B cells, monocytes, and T cell leukemias. CD122 can also be detected during early NK cell commitment. IL-2R beta associates with the IL-2R alpha (CD25) or IL-15R alpha and common gamma chain (CD132) to form the high affinity IL-2 and IL-15 receptors, respectively. By itself, this membrane receptor can bind IL-2, but with intermediate affinity. The binding of IL-2 results in internalization of the cytokine, tyrosine phosphorylation of IL-2Rβ, and activation of signaling cascades such as the JAK/STAT pathway.
Description & Specifications

Specifications

CD122
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TU27
Interleukin-2 Receptor beta, IL-2Rb, IL-2Rp75
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test