missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD11c Mouse, APC-eFluor 780, Clone: BU15, eBioscience™

Mouse Monoclonal Antibody

$224.40 - $472.56

Specifications

Antigen CD11c
Clone BU15
Host Species Murine
Gene Alias Integrin alpha-X, p150, CR4, Integrin aX, ITGAX
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124010 Life Technologies
LS47012842
100 Tests Each for $472.56
501129279 Life Technologies
LS47012841
25 Tests Each for $224.40
Specifications

Description

The BU15 monoclonal antibody reacts with canine, human, and non-human primate CD11c, the 150kDa integrin aX chain. CD11c non-covalently associates with beta2 integrin to form the CD11c/CD18 heterodimer. This complex is expressed on monocytes, granulocytes, macrophages, NK, dendritic cells, and subset of T and B lymphocytes. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions. CD11c also binds complement fragment iC3b to mediate phagocytosis of opsonized particles.

The BU15 antibody has been reported to crossreact to cynomologus monkey. Moreover, this antibody has been reported to exhibit blocking activity. Crossblocking studies suggest that BU15 recognizes a different epitope from clone 3.9.
Description & Specifications

Specifications

CD11c
Murine
Flow Cytometry
APC-eFluor 780
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.125μg)/test
BU15
Integrin alpha-X, p150, CR4, Integrin aX, ITGAX
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer