Please login to your online account to display your discounted pricing

CD11c Mouse anti-Human, PE-Cy7, Clone: 3.9, eBioscience™

Mouse Monoclonal Antibody

$220.40 - $456.75

Specifications

Antigen CD11c
Clone 3.9
Host Species Mouse
Gene Alias Integrin alpha X, Integrin aX, ITGAX, p150/95, leu M5 alpha
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015457

 
life technologies
LS25011641
25 tests Each for $220.40

501129682

 
life technologies
LS25011642
100 tests Each for $456.75
Description & Specifications

Specifications

Antigen CD11c
Clone 3.9
Host Species Mouse
Gene Alias Integrin alpha X, Integrin aX, ITGAX, p150/95, leu M5 alpha
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (1.0μg)/test

The 3.9 monoclonal antibody reacts with human CD11c, the 150kDa integrin alpha X chain. CD11c non-covalently associates with beta2 integrin to form the CD11c/CD18 heterodimer. This complex is expressed on monocytes, granulocytes, macrophages, NK, dendritic cells and subset of T and B lymphocytes. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions.