Please login to your online account to display your discounted pricing

CD11c Mouse anti-Human, PE-Cy5, Clone: 3.9, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $404.55

Specifications

Antigen CD11c
Clone 3.9
Host Species Murine
Gene Alias Integrin alpha X, Integrin aX, ITGAX
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122907

 
life technologies
LS15011641
25 Tests Each for $158.05

501122908

 
life technologies
LS15011642
100 Tests Each for $404.55
Description & Specifications

Specifications

Antigen CD11c
Clone 3.9
Host Species Murine
Gene Alias Integrin alpha X, Integrin aX, ITGAX
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

The 3.9 monoclonal antibody reacts with human CD11c, the 150kDa integrin alpha X chain. CD11c non-covalently associates with beta2 integrin to form the CD11c/CD18 heterodimer. This complex is expressed on monocytes, granulocytes, macrophages, NK, dendritic cells and subset of T and B lymphocytes. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions.