missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD11c Mouse anti-Human, PE-Cy5.5, Clone: 3.9, eBioscience™

Mouse Monoclonal Antibody

$161.04 - $394.68

Specifications

Antigen CD11c
Clone 3.9
Host Species Murine
Gene Alias Integrin alpha X, Integrin aX, ITGAX
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123541 Life Technologies
LS35011641
25 Tests Each for $161.04
501123542 Life Technologies
LS35011642
100 Tests Each for $394.68
Specifications

Description

The 3.9 monoclonal antibody reacts with human CD11c, the 150kDa integrin alpha X chain. CD11c non-covalently associates with beta2 integrin to form the CD11c/CD18 heterodimer. This complex is expressed on monocytes, granulocytes, macrophages, NK, dendritic cells and subset of T and B lymphocytes. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions.
Description & Specifications

Specifications

CD11c
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
3.9
Integrin alpha X, Integrin aX, ITGAX
Flow Cytometry
PE-Cy5.5
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (1.0μg)/test