Please login to your online account to display your discounted pricing

CD11c Mouse anti-Human, Biotin, Clone: 3.9, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $259.55

Specifications

Antigen CD11c
Clone 3.9
Host Species Murine
Gene Alias Integrin alpha X, Integrin aX, ITGAX, p150/95, leu M5 alpha
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011436

 
life technologies
LS13011680
25μg Each for $114.55

5011437

 
life technologies
LS13011682
100μg Each for $259.55
Description & Specifications

Specifications

Antigen CD11c
Clone 3.9
Host Species Murine
Gene Alias Integrin alpha X, Integrin aX, ITGAX, p150/95, leu M5 alpha
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 3.9 monoclonal antibody reacts with human CD11c, the 150kDa integrin αX chain. CD11c non-covalently associates with β2 integrin to form the CD11c/CD18 heterodimer. This complex is expressed on monocytes, granulocytes, macrophages, NK, dendritic cells and subset of T and B lymphocytes. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions.