missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD11c Armenian Hamster anti-Mouse, PE-Cy5, Clone: N418, eBioscience™

Armenian Hamster Monoclonal Antibody

$249.40 - $427.75

Specifications

Antigen CD11c
Clone N418
Host Species Armenian Hamster
Gene Alias Integrin alpha X, Integrin aX, ITGAX, p150/95, leu M5 alpha
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014016 Life Technologies
LS15011481
50μg Each for $249.40
5014017 Life Technologies
LS15011482
100μg Each for $427.75
Specifications

Description

The N418 monoclonal antibody reacts with mouse CD11c, the integrin alpha X. CD11c non-covalently associates with beta 2 integrin to form the CD11c/CD18 heterodimer. CD11c is expressed by dendritic cells, a subset of Intestinal Intraepithelial Lymphocytes (IEL) and some activated T cells. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions. N418 binds to CD11c on splenic dendritic cells in the T-dependent areas of mouse spleen and precipitates a 150, 90kDa heterodimer.
Description & Specifications

Specifications

CD11c
Armenian Hamster
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
N418
Integrin alpha X, Integrin aX, ITGAX, p150/95, leu M5 alpha
Flow Cytometry
PE-Cy5
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL