Please login to your online account to display your discounted pricing

CD11c Armenian Hamster anti-Mouse, Alexa Fluor 700, Clone: N418, eBioscience™

Armenian Hamster Monoclonal Antibody

$143.55 - $317.55

Specifications

Antigen CD11c
Clone N418
Host Species Armenian Hamster
Gene Alias Integrin alpha X, Integrin aX, ITGAX, p150/95, leu M5 alpha
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016868

 
affymetrix
56011480
25μg Each for $143.55

5016869

 
affymetrix
56011482
100μg Each for $317.55
Description & Specifications

Specifications

Antigen CD11c
Clone N418
Host Species Armenian Hamster
Gene Alias Integrin alpha X, Integrin aX, ITGAX, p150/95, leu M5 alpha
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Alexa Fluor 700
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The N418 monoclonal antibody reacts with mouse CD11c, the integrin αX. CD11c non-covalently associates with β2 integrin to form the CD11c/CD18 heterodimer. CD11c is expressed by dendritic cells, a subset of Intestinal Intraepithelial Lymphocytes (IEL) and some activated T cells. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions. N418 binds to CD11c on splenic dendritic cells in the T-dependent areas of mouse spleen and precipitates a 150, 90kDa heterodimer.