Please login to your online account to display your discounted pricing

CD11b Rat anti-Mouse, PE, Clone: M1/70, eBioscience™

Rat Monoclonal Antibody

$129.05 - $651.05

Specifications

Antigen CD11b
Clone M1/70
Host Species Rat
Gene Alias Integrin alpha M
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010214

 
life technologies
LS12011283
200μg Each for $404.55

5010215

 
life technologies
LS12011285
500μg Each for $651.05

501129669

 
life technologies
LS12011282
100μg Each for $201.55

501129671

 
life technologies
LS12011281
50μg Each for $129.05
Description & Specifications

Specifications

Antigen CD11b
Clone M1/70
Host Species Rat
Gene Alias Integrin alpha M
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The M1/70 monoclonal antibody reacts with mouse CD11b, the 165-170kDa integrin αM. CD11b non-covalently associates with CD18 to form αMβ2 integrin (Mac-1) and binds to CD54 (ICAM-1), C3bi, and fibrinogen. Mac-1 is expressed by macrophages, NK cells, granulocytes, activated lymphocytes and mouse B-1 cells in the peritoneal cavity. M1/70 is also cross-reactive to human CD11b, and can be used for the detection of this antigen on human peripheral blood monocytes, granulocytes, and a subset of NK cells. Through interactions with its ligands, CD11b participates in adhesive cell interactions.