missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD11b Rat anti-Mouse, FITC, Clone: M1/70, eBioscience™

Rat Monoclonal Antibody

$91.08 - $658.68

Specifications

Antigen CD11b
Clone M1/70
Host Species Rat
Gene Alias Integrin alpha M
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122037 Life Technologies
LS11011241
25 Tests Each for $104.28
509434 Life Technologies
LS11011281
50μg Each for $91.08
509436 Life Technologies
LS11011285
500μg Each for $381.48
509437 Life Technologies
LS11011286
1mg Each for $658.68
501129521 Life Technologies
LS11011282
100μg Each for $157.08
Specifications

Description

The M1/70 monoclonal antibody reacts with mouse CD11b, the 165-170kDa integrin αM. CD11b non-covalently associates with CD18 to form αMβ2 integrin (Mac-1) and binds to CD54 (ICAM-1), C3bi, and fibrinogen. Mac-1 is expressed by macrophages, NK cells, granulocytes, activated lymphocytes and mouse B-1 cells in the peritoneal cavity. M1/70 is also cross-reactive to human CD11b, and can be used for the detection of this antigen on human peripheral blood monocytes, granulocytes, and a subset of NK cells. Through interactions with its ligands, CD11b participates in adhesive cell interactions.
Description & Specifications

Specifications

CD11b
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
M1/70
Integrin alpha M
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL; 5μL (0.25μg)/test