missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD11b Rat anti-Mouse, Biotin, Clone: M1/70, eBioscience™

Rat Monoclonal Antibody

$100.05 - $694.55

Specifications

Antigen CD11b
Clone M1/70
Host Species Rat
Gene Alias Integrin alpha M, ITGAM, Mac-1 alpha (Mac1A), Complement Receptor 3 alpha (CR3A)
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122503 Life Technologies
LS13011281
50μg Each for $100.05
5011428 Life Technologies
LS13011282
100μg Each for $187.05
5011429 Life Technologies
LS13011285
500μg Each for $361.05
5011430 Life Technologies
LS13011286
1mg Each for $694.55
Specifications

Description

The M1/70 monoclonal antibody reacts with mouse CD11b, the 165-170kDa integrin αM. CD11b non-covalently associates with CD18 to form αMβ2 integrin (Mac-1) and binds to CD54 (ICAM-1), C3bi, and fibrinogen. Mac-1 is expressed by macrophages, NK cells, granulocytes, activated lymphocytes and mouse B-1 cells in the peritoneal cavity. M1/70 is also cross-reactive to human CD11b, and can be used for the detection of this antigen on human peripheral blood monocytes, granulocytes, and a subset of NK cells. Through interactions with its ligands, CD11b participates in adhesive cell interactions.
Description & Specifications

Specifications

CD11b
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
M1/70
Integrin alpha M, ITGAM, Mac-1 alpha (Mac1A), Complement Receptor 3 alpha (CR3A)
Flow Cytometry,Immunohistochemistry (Frozen)
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL