Please login to your online account to display your discounted pricing

CD11b Rat anti-Mouse, Alexa Fluor 700, Clone: M1/70, eBioscience™

Rat Monoclonal Antibody

$172.55 - $317.55

Specifications

Antigen CD11b
Clone M1/70
Host Species Rat
Gene Alias Integrin alpha M
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5016864

 
affymetrix
56011280
25μg Each for $172.55

5016865

 
affymetrix
56011282
100μg Each for $317.55
Description & Specifications

Specifications

Antigen CD11b
Clone M1/70
Host Species Rat
Gene Alias Integrin alpha M
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Alexa Fluor 700
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The M1/70 monoclonal antibody reacts with mouse CD11b, the 165-170kDa integrin αM. CD11b non-covalently associates with CD18 to form αMβ2 integrin (Mac-1) and binds to CD54 (ICAM-1), C3bi, and fibrinogen. Mac-1 is expressed by macrophages, NK cells, granulocytes, activated lymphocytes and mouse B-1 cells in the peritoneal cavity. M1/70 is also cross-reactive to human CD11b, and can be used for the detection of this antigen on human peripheral blood monocytes, granulocytes, and a subset of NK cells. Through interactions with its ligands, CD11b participates in adhesive cell interactions.