missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD11b Mouse anti-Human, PerCP-eFluor 710, Clone: ICRF44, eBioscience™

Mouse Monoclonal Antibody

$145.20 - $389.40

Specifications

Antigen CD11b
Clone ICRF44
Host Species Murine
Gene Alias Integrin alpha M, ITGAM, Mac-1 alpha (Mac1A), Complement Receptor 3 alpha (CR3A)
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015917 Life Technologies
LS46011841
25 Tests Each for $145.20
5015918 Life Technologies
LS46011842
100 Tests Each for $389.40
Specifications

Description

The ICRF44 monoclonal antibody reacts with human CD11b, a 165kDa adhesion molecule. CD11b associated with integrin β2 (CD18) is expressed on the surface of monocytes, granulocytes, activated lymphocytes and a subset of NK cells. CD11b is a receptor for intercellular adhesion molecule family members CD54, CD102 and CD50 as well as for iC3b. These adhesions are crucial in cell-cell and cell-matrix interactions.
Description & Specifications

Specifications

CD11b
Murine
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
ICRF44
Integrin alpha M, ITGAM, Mac-1 alpha (Mac1A), Complement Receptor 3 alpha (CR3A)
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer