Please login to your online account to display your discounted pricing

CD11b Mouse anti-Human, FITC, Clone: ICRF44, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $274.05

Specifications

Antigen CD11b
Clone ICRF44
Host Species Mouse
Gene Alias Integrin alpha M, Mac-1 alpha, Complement Receptor 3 alpha
Species Reactivity Baboon, Chimpanzee, Cynomolgus, Human, Rhesus Ma
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509445

 
life technologies
LS11011841
25 tests Each for $129.05

509446

 
life technologies
LS11011842
100 tests Each for $274.05
Description & Specifications

Specifications

Antigen CD11b
Clone ICRF44
Host Species Mouse
Gene Alias Integrin alpha M, Mac-1 alpha, Complement Receptor 3 alpha
Species Reactivity Baboon, Chimpanzee, Cynomolgus, Human, Rhesus Ma
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The ICRF44 monoclonal antibody reacts with human CD11b, a 165kDa adhesion molecule. CD11b associated with integrin beta2 (CD18) is expressed on the surface of monocytes, granulocytes, activated lymphocytes and a subset of NK cells. CD11b is a receptor for intercellular adhesion molecule family members CD54, CD102 and CD50 as well as for iC3b. These adhesions are crucial in cell-cell and cell-matrix interactions.