missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD11b Mouse anti-Human, APC-eFluor 780, Clone: ICRF44, eBioscience™

Mouse Monoclonal Antibody

$183.48 - $381.48

Specifications

Antigen CD11b
Clone ICRF44
Host Species Murine
Gene Alias Integrin alpha M, Mac-1 alpha, Complement Receptor 3 alpha
Species Reactivity Baboon, Chimpanzee, Cynomolgus, Human, Rhesus Ma
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016145 Life Technologies
LS47011841
25 Tests Each for $183.48
5016146 Life Technologies
LS47011842
100 Tests Each for $381.48
Specifications

Description

The ICRF44 monoclonal antibody reacts with human CD11b, a 165kDa adhesion molecule. CD11b associated with integrin beta2 (CD18) is expressed on the surface of monocytes, granulocytes, activated lymphocytes and a subset of NK cells. CD11b is a receptor for intercellular adhesion molecule family members CD54, CD102 and CD50 as well as for iC3b. These adhesions are crucial in cell-cell and cell-matrix interactions.
Description & Specifications

Specifications

CD11b
Murine
Baboon, Chimpanzee, Cynomolgus, Human, Rhesus Ma
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
ICRF44
Integrin alpha M, Mac-1 alpha, Complement Receptor 3 alpha
Flow Cytometry
APC-eFluor 780
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.25μg)/test