missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD11b Mouse anti-Human, Alexa Fluor 700, Clone: CBRM1/5, eBioscience™

Mouse Monoclonal Antibody

$274.05 - $462.55

Specifications

Antigen CD11b
Clone CBRM1/5
Host Species Murine
Gene Alias Integrin alpha M, Mac-1 alpha, Complement Receptor 3 alpha (CR3A)
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016866 Life Technologies
LS56011341
25 Tests Each for $274.05
5016867 Life Technologies
LS56011342
100 Tests Each for $462.55
Specifications

Description

The CBRM1/5 monoclonal antibody reacts with an activation-specific epitope of human Mac-1. CBRM1/5 binds a subset of Mac-1 molecules on neutrophils and monocytes after stimulation with chemoattractants or phorbol esters but does not recognize Mac-1 on resting myeloid cells. Through interactions with its ligands, Mac-1 participates in adhesive cell interactions. The epitope recognized by this mAb localizes to the I domain on the α chain of Mac-1 very close to the ligand binding site in a region that is widely exposed. CBRM1/5 blocks Mac-1 dependent adhesion to fibrinogen and ICAM-1 and inhibits chemoattractant-stimulated adhesion of eosinophils to the Intercellular Adhesion Molecule-1 (ICAM-1). It should be noted that low level activation may occur during processing of freshly drawn blood. Therefore the CBRM1.5 antibody may exhibit some binding to Mac-1 in these unstimulated samples. However, higher levels of Mac-1 expression are observed in activated samples when compared to unstimulated cells.
Description & Specifications

Specifications

CD11b
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CBRM1/5
Integrin alpha M, Mac-1 alpha, Complement Receptor 3 alpha (CR3A)
Flow Cytometry
Alexa Fluor 700
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test