missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD11b/c, PE, clone: OX42, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS12011080

 View more versions of this product

Catalog No. 501129365



Description

Description

The principal use of this antibody is in the study of functional heterogeneity of macrophages; it may be used to follow macrophage differentiation and to investigate the function of the CD11b/c equivalent antigen.

Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. CD11b is implicated in various adhesive interactions of monocytes, macrophages and granulocytes as well as in mediating the uptake of complement coated particles. It is identical to CR3, the receptor for the iC3b fragment of the third complement component. CD11b is also a receptor for fibrogen, factor X and ICAM1. It recognizes P1 and P2 peptides of fibrogen gamma chain. The Mac1 CD11b antigen is present on macrophages, granulocytes, natural killer cells, blood monocytes. CD11b is expressed on 8% spleen cells, 44% bone marrow cells and less than 1% thymocytes and is commonly used as a microgial marker in nervous tissue. CD11c encodes the integrin alpha X chain protein. This protein combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as inactivated-C3b (iC3b) receptor 4 (CR4). The alpha X beta 2 complex seems to overlap the properties of the alpha M beta 2 integrin in the adherence of neutrophils and monocytes to stimulated endothelium cells, and in the phagocytosis of complement coated particles. Mutations affecting the CD11b/c gene lead to systemic lupus erythematosus 6.
Specifications

Specifications

CD11b/c
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Liquid
Itgam, Itgax
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
25021, 499271
Flow Cytometry
OX42
PE
Itgax
Itgax, Itgam, CD11B, CR3A, MAC-1, MAC1A, MGC117044, MO1A, SLEB6, CD11bc
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Rat
Documents

For Research Use Only.