missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD11b/c Mouse anti-Rat, PerCP-eFluor 710, Clone: OX42, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $328.68

Specifications

Antigen CD11b/c
Clone OX42
Host Species Murine
Gene Alias C3bi, Mac-1, Integrin alpha M, CR4
Species Reactivity Rat
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245849 Life Technologies
LS46011080
25μg Each for $157.08
50245850 Life Technologies
LS46011082
100μg Each for $328.68
Specifications

Description

This OX42 monoclonal antibody reacts with rat CD11b and CD11c. CD11b, also known as integrin alpha M or Mac-1, is a component of complement receptor 3 (CR3). CD11c, also known as integrin alpha X, is a component of complement receptor 4 (CR4). CD11b and CD11c are expressed on immune cells such as macrophages, monocytes, granulocytes, and dendritic cells. OX42 has also been shown to detect microglia in the brain, as well as cells of the liver and epidermis.

The OX42 antibody has been reported to inhibit complement-mediated rosette formation by granulocytes and macrophages. Moreover, studies demonstrate that OX42 can inhibit granulocyte aggregation.
Description & Specifications

Specifications

CD11b/c
Murine
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
OX42
C3bi, Mac-1, Integrin alpha M, CR4
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL