Please login to your online account to display your discounted pricing

CD11b/c Mouse anti-Rat, eFluor 660, Clone: OX42, eBioscience™

Mouse Monoclonal Antibody

$137.75 - $311.75

Specifications

Antigen CD11b/c
Clone OX42
Host Species Mouse
Gene Alias C3bi, Mac-1, Integrin alpha M, CR4
Species Reactivity Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124250

 
life technologies
LS50011080
25μg Each for $137.75

501124251

 
life technologies
LS50011082
100μg Each for $311.75
Description & Specifications

Specifications

Antigen CD11b/c
Clone OX42
Host Species Mouse
Gene Alias C3bi, Mac-1, Integrin alpha M, CR4
Species Reactivity Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This OX42 monoclonal antibody reacts with rat CD11b and CD11c. CD11b, also known as integrin alpha M or Mac-1, is a component of complement receptor 3 (CR3). CD11c, also known as integrin alpha X, is a component of complement receptor 4 (CR4). CD11b and CD11c are expressed on immune cells such as macrophages, monocytes, granulocytes, and dendritic cells. OX42 has also been shown to detect microglia in the brain, as well as cells of the liver and epidermis.

The OX42 antibody has been reported to inhibit complement-mediated rosette formation by granulocytes and macrophages. Moreover, studies demonstrate that OX42 can inhibit granulocyte aggregation.