Please login to your online account to display your discounted pricing

CD11a Rat anti-Mouse, eFluor 450, Clone: M17/4, eBioscience™

Rat Monoclonal Antibody

$129.05 - $274.05

Specifications

Antigen CD11a
Clone M17/4
Host Species Rat
Gene Alias Integrin alpha L, Lymphocyte Function-Associated Antigen
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124089

 
life technologies
LS48011180
25μg Each for $129.05

501129328

 
life technologies
LS48011182
100μg Each for $274.05
Description & Specifications

Specifications

Antigen CD11a
Clone M17/4
Host Species Rat
Gene Alias Integrin alpha L, Lymphocyte Function-Associated Antigen
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The M17/4 monoclonal antibody reacts with mouse CD11a, the 180kDa integrin alpha L, also known as the lymphocyte function associated antigen-1 (LFA-1) alpha chain. LFA-1, formed by non-covalent association of CD11a with CD18 (integrin beta 2), serves as an important adhesion molecule involved in lymphocyte and granulocyte function. CD54 (ICAM-1), CD102 (ICAM-2), and CD50 (ICAM-3) are ligands for LFA-1. CD11a is expressed by all leukocytes.