missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD11a Rat anti-Mouse, Clone: M17/4, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$104.28 - $368.28

Specifications

Antigen CD11a
Clone M17/4
Host Species Rat
Gene Alias Integrin alpha L, Lymphocyte Function-Associated Antigen
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014221 Life Technologies
LS16011181
50μg Each for $104.28
5014222 Life Technologies
LS16011182
100μg Each for $196.68
5014223 Life Technologies
LS16011185
500μg Each for $368.28
Specifications

Description

The M17/4 monoclonal antibody reacts with mouse CD11a, the 180kDa integrin alpha L, also known as the lymphocyte function associated antigen-1 (LFA-1) alpha chain. LFA-1, formed by non-covalent association of CD11a with CD18 (integrin beta 2), serves as an important adhesion molecule involved in lymphocyte and granulocyte function. CD54 (ICAM-1), CD102 (ICAM-2), and CD50 (ICAM-3) are ligands for LFA-1. CD11a is expressed by all leukocytes.
Description & Specifications

Specifications

CD11a
Rat
Flow Cytometry
Unlabeled
Store at 2-8°C.
Monoclonal
M17/4
Integrin alpha L, Lymphocyte Function-Associated Antigen
RUO
Purified
Primary
aqueous buffer, no sodium azide