Please login to your online account to display your discounted pricing

CD11a Mouse anti-Human, Clone: HI111, eBioscience™

Mouse Monoclonal Antibody

$71.05 - $158.05

Specifications

Antigen CD11a
Clone HI111
Host Species Mouse
Gene Alias Integrin alpha L, ITGAL, Lymphocyte Function-Associated Antigen, Type 1 alpha (LFA-1 alpha), LFA1A
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012428

 
life technologies
LS14011982
100μg Each for $158.05

5012427

 
life technologies
LS14011980
25μg Each for $71.05
Description & Specifications

Specifications

Antigen CD11a
Clone HI111
Host Species Mouse
Gene Alias Integrin alpha L, ITGAL, Lymphocyte Function-Associated Antigen, Type 1 alpha (LFA-1 alpha), LFA1A
Species Reactivity Human
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The HI111 monoclonal antibody reacts with human CD11a, the 180kDa integrin αL, also known as the lymphocyte function associated antigen-1 (LFA-1) α chain. LFA-1, formed by non-covalent association of CD11a with CD18 (integrin β2), serves as an important adhesion molecule involved in lymphocyte and granulocyte function. CD54 (ICAM-1), CD102 (ICAM-2), and CD50 (ICAM-3) are ligands for LFA-1. CD11a is expressed by all leukocytes.