Please login to your online account to display your discounted pricing

CD119 (IFN gamma Receptor 1) Mouse anti-Human, Biotin, Clone: GIR-208 (GIR 208), eBioscience™

Mouse Monoclonal Antibody

$71.05 - $172.55

Specifications

Antigen CD119 (IFN gamma Receptor 1)
Clone GIR-208 (GIR 208)
Host Species Murine
Gene Alias IFNgR1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122522

 
life technologies
LS13119980
25μg Each for $71.05

501122523

 
life technologies
LS13119982
100μg Each for $172.55
Description & Specifications

Specifications

Antigen CD119 (IFN gamma Receptor 1)
Clone GIR-208 (GIR 208)
Host Species Murine
Gene Alias IFNgR1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody reacts with human CD119 also known as the IFN gamma receptor 1 (alpha chain). IFN gamma R1 is found in a complex with IFN gamma receptor beta and recognizes with high affinity IFN gamma. Expression is ubiquitous but low, although some cell types, such as monocytes express higher levels.

The GIR-208 monoclonal antibody is blocked from binding when IFN gamma is bound to the receptor.