Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD117/c-Kit/SCF-Receptor, Rabbit Polyclonal Antibody

Optimize staining of formalin-fixed, paraffin-embedded tissues with Thermo Scientific™ CD117/c-Kit/SCF-Receptor, Rabbit Polyclonal Antibody.

$257.82 - $1,298.76

Specifications

Antigen CD117/c-Kit/SCF-Receptor
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

RB9038P

 
thermo scientific™ lab vision™
RB9038P
1mL Each for $1,298.76

RB9038P0

 
thermo scientific™ lab vision™
RB9038P0
100µL Each for $257.82

RB9038P1

 
thermo scientific™ lab vision™
RB9038P1
500µL Each for $766.24

RB9038R7

 
thermo scientific™ lab vision™
RB9038R7
7mL Each for $631.39
Description & Specifications

Specifications

Antigen CD117/c-Kit/SCF-Receptor
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Host Species Rabbit
Regulatory Status IVD
Type Primary
Immunogen A synthetic peptide from the cytoplasmic domain of human CD117 / c-kit protein

Optimize staining of formalin-fixed, paraffin-embedded tissues with Thermo Scientific™ CD117/c-Kit/SCF-Receptor, Rabbit Polyclonal Antibody, one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Recognizes a protein of 145kDa, which is identified as CD117/p145kit. This rabbit polyclonal antibody does not interfere with the binding of SCF to c-kit. It precipitates both the unoccupied as well as the occupied form of c-kit. The binding of the stem cell factor (SCF) to the c-kit-encoded receptor tyrosine kinase (Type III) stimulates a variety of biochemical responses that culminate in cellular proliferation, migration, or survival. C-kit plays an important role in hematopoiesis, melanogenesis, and gametogenesis. This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Dog. Others not tested

Epitope: Cytoplasmic domain

Immunogen: A synthetic peptide from the cytoplasmic domain of human CD117/c-kit protein

Molecular Weight: 145 kDa

Positive Control: Stromal tumor, skin or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)