missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD117/c-Kit/SCF-Receptor Ab-6, Rabbit Polyclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD117/c-Kit/SCF-Receptor Ab-6, Rabbit Polyclonal Antibody.

$278.67 - $1,406.16

Specifications

Antigen CD117/c-kit/SCF-Receptor Ab-6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Format Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Format Quantity Price Quantity  
RB1518P Thermo Scientific™ Lab Vision™
RB1518P
4mg/mL Concentrated 1mL Each for $1,286.22
RB1518P0 Thermo Scientific
RB1518P0
4mg/mL Purified with BSA and azide 100μL Each for $278.67
RB1518P1 Thermo Scientific
RB1518P1
4mg/mL Purified with BSA and azide 500μL Each for $776.88
RB1518P1ABX Thermo Scientific™ Lab Vision™
RB1518P1ABX
4mg/mL Purified without BSA and azide 500μL Each for $846.04
RB1518PABX Thermo Scientific™ Lab Vision™
RB1518PABX
4mg/mL Purified without BSA and azide 1mL Each for $1,406.16
RB1518R7 Thermo Scientific™ Lab Vision™
RB1518R7
---- Prediluted, Ready to Use 7mL Each for $680.94
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD117/c-Kit/SCF-Receptor Ab-6, Rabbit Polyclonal Antibody.

c-kit is a transmembrane receptor tyrosine kinase. It binds to stem cell factor (SCF). It is expressed in many tissues and cells. C-kit/SCf is involved in development of several lineages of stem cells, such as germ cells, neural crest-derived melanocytes and hematopoietic precursor cells Antibody to CD117/c-kit may be helpful in the histopathological identification of gastrointestinal stromal tumors (GIST).

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human CD117/c-kit protein

Molecular Weight: 145kDa

Positive Control: Tonsil, GI Stromal Tumor (GIST)

Cellular Localization: Membrane/cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

CD117/c-kit/SCF-Receptor Ab-6
Polyclonal
Human
RUO
A synthetic peptide from C-terminus of human CD117/c-kit protein
Immunohistochemistry (Paraffin)
Unlabeled
Rabbit
Primary
IgG