Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD117/c-Kit/SCF-Receptor Ab-6, Rabbit Polyclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD117/c-Kit/SCF-Receptor Ab-6, Rabbit Polyclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  RB1518P1ABX

 View more versions of this product

Catalog No. RB1518P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Isotype IgG
Quantity 500µL
Immunogen A synthetic peptide from C-terminus of human CD117/c-kit protein
Cross Reactivity Human
Host Species Rabbit
Clone Polyclonal
Format Purified w/o BSA, azide
Regulatory Status RUO
Antigen CD117/c-Kit/SCF-Receptor Ab-6
Classification Polyclonal
Conjugate Unlabeled
Concentration 4mg/mL
Type Primary

Ensure accurate, reproducible results in immunohistochemistry applications with Thermo Scientific™ CD117/c-Kit/SCF-Receptor Ab-6, Rabbit Polyclonal Antibody.

c-kit is a transmembrane receptor tyrosine kinase. It binds to stem cell factor (SCF). It is expressed in many tissues and cells. C-kit/SCf is involved in development of several lineages of stem cells, such as germ cells, neural crest-derived melanocytes and hematopoietic precursor cells Antibody to CD117/c-kit may be helpful in the histopathological identification of gastrointestinal stromal tumors (GIST).

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: C-terminal

Immunogen: A synthetic peptide from C-terminus of human CD117/c-kit protein

Molecular Weight: 145kDa

Positive Control: Tonsil, GI Stromal Tumor (GIST)

Cellular Localization: Membrane/cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin only)

Note:

USA: RUO; Int'l: RUO