Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD117/c-Kit/SCF-Receptor Ab-3, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments.

$242.55 - $1,145.08

Specifications

Antigen CD117/c-Kit/SCF-Receptor Ab-3
Clone K45
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunofluorescence
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS289P

 
thermo scientific™ lab vision™
MS289P
200μg/mL 1mL Each for $1,059.76

MS289P0

 
thermo scientific™ lab vision™
MS289P0
200μg/mL 100µL Each for $242.55

MS289P1

 
thermo scientific™ lab vision™
MS289P1
200μg/mL 500µL Each for $713.22

MS289P1ABX

 
thermo scientific™ lab vision™
MS289P1ABX
1mg/mL 100µL Each for $785.10

MS289PABX

 
thermo scientific™ lab vision™
MS289PABX
1mg/mL 200µL Each for $1,145.08
Description & Specifications

Specifications

Antigen CD117/c-Kit/SCF-Receptor Ab-3
Clone K45
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Type Primary
Immunogen Recombinant human extracellular c-kit fragment
Isotype IgG2a

Obtain accurate, reproducible results in immunohistochemistry and immunofluorescence experiments with Thermo Scientific™ CD117/c-Kit/SCF-Receptor Ab-3, Mouse Monoclonal Antibody.

c-kit is a transmembrane receptor tyrosine kinase. It binds to stem cell factor (SCF). It is expressed in many tissues and cells. C-kit/SCf is involved in development of several lineages of stem cells, such as germ cells, neural crest-derived melanocytes and hematopoietic precursor cells. Ab-3 does not interfere with the binding of SCF to c-kit

Host Species: Mouse

Clone: K45

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Extracellular second N-terminus Ig-like domain of c-kit

Immunogen: Recombinant human extracellular c-kit fragment

Molecular Weight: 145kDa

Positive Control: Skin, GI Stromal Tumor (GIST)

Cellular Localization: Membrane/cytoplasmic

Recommended for:

  • Immunohistochemistry (Frozen or fixed in methacarn or Stieve's and embedded in paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO