missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD117 (c-kit) Rat anti-Mouse, PerCP-eFluor 710, Clone: 2B8, eBioscience™

Rat Monoclonal Antibody

$147.84 - $473.88

Specifications

Antigen CD117 (c-kit)
Clone 2B8
Host Species Rat
Gene Alias Steel Factor Receptor
Species Reactivity Mouse, Pig
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016003 Life Technologies
LS46117180
25μg Each for $147.84
5016004 Life Technologies
LS46117182
100μg Each for $473.88
Specifications

Description

The 2B8 monoclonal antibody reacts with mouse CD117, also known as c-Kit receptor, Steel factor receptor, and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145kDa molecule is expressed by a majority of hematopoietic progenitor cells characterized in the mouse bone marrow as a small subset of cells positive for Sca-1 and Thy1 (Thy1 low) and negative for lineage markers. The interaction of the mouse c-Kit receptor and steel factor promotes the proliferation and differentiation of hematopoietic progenitor cells. CD117 is also expressed by mast cells and plays a role in signaling and activation of these cells.
Description & Specifications

Specifications

CD117 (c-kit)
Rat
Mouse, Pig
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
2B8
Steel Factor Receptor
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL