Please login to your online account to display your discounted pricing

CD117 (c-kit) Rat anti-Mouse, APC, Clone: 2B8, eBioscience™

Rat Monoclonal Antibody

$220.40 - $516.20

Specifications

Antigen CD117 (c-kit)
Clone 2B8
Host Species Rat
Gene Alias Steel Factor Receptor
Species Reactivity Mouse, Pig
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015055

 
life technologies
LS17117183
200μg Each for $516.20

501129173

 
life technologies
LS17117182
100μg Each for $288.55

501129382

 
life technologies
LS17117181
50μg Each for $220.40
Description & Specifications

Specifications

Antigen CD117 (c-kit)
Clone 2B8
Host Species Rat
Gene Alias Steel Factor Receptor
Species Reactivity Mouse, Pig
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 2B8 monoclonal antibody reacts with mouse CD117, also known as c-Kit receptor, Steel factor receptor, and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145kDa molecule is expressed by a majority of hematopoietic progenitor cells characterized in the mouse bone marrow as a small subset of cells positive for Sca-1 and Thy1 (Thy1 low) and negative for lineage markers. The interaction of the mouse c-Kit receptor and steel factor promotes the proliferation and differentiation of hematopoietic progenitor cells. CD117 is also expressed by mast cells and plays a role in signaling and activation of these cells.