missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD117 (c-kit) Mouse anti-Human, FITC, Clone: 104D2, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $317.55

Specifications

Antigen CD117 (c-kit)
Clone 104D2
Host Species Murine
Gene Alias SCFR, Steel Factor Receptor
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122090 Life Technologies
LS11117841
25 Tests Each for $129.05
501122091 Life Technologies
LS11117842
100 Tests Each for $317.55
Specifications

Description

The 104D2 monoclonal antibody reacts with human CD117, also known as c-Kit, Steel factor receptor and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145kDa molecule is expressed by hematopoietic progenitor cell subsets and mast cells. The interaction of c-Kit and Steel factor promotes proliferation and differentiation of hematopoietic progenitor cells and mast cell differentiation. CD117 is also expressed by melanocytes and plays a role in signaling and activation of these cells.
Description & Specifications

Specifications

CD117 (c-kit)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
104D2
SCFR, Steel Factor Receptor
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test