missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD117 (c-kit) Mouse anti-Human, eFluor 450, Clone: 104D2, eBioscience™

Mouse Monoclonal Antibody

$209.88 - $460.68

Specifications

Antigen CD117 (c-kit)
Clone 104D2
Host Species Murine
Gene Alias SCFR, Steel Factor Receptor
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50246000 Life Technologies
LS48117841
25 Tests Each for $209.88
50246001 Life Technologies
LS48117842
100 Tests Each for $460.68
Specifications

Description

The 104D2 monoclonal antibody reacts with human CD117, also known as c-Kit, Steel factor receptor and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145kDa molecule is expressed by hematopoietic progenitor cell subsets and mast cells. The interaction of c-Kit and Steel factor promotes proliferation and differentiation of hematopoietic progenitor cells and mast cell differentiation. CD117 is also expressed by melanocytes and plays a role in signaling and activation of these cells.
Description & Specifications

Specifications

CD117 (c-kit)
Murine
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
104D2
SCFR, Steel Factor Receptor
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer