Please login to your online account to display your discounted pricing

CD116 Mouse anti-Human, PE, Clone: 4H1, eBioscience™

Mouse Monoclonal Antibody

$288.55 - $288.55

Specifications

Antigen CD116
Clone 4H1
Host Species Mouse
Gene Alias GM-CSF Receptor alpha, CSF2RA
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010567

 
life technologies
LS12116942
100 tests Each for $288.55
Description & Specifications

Specifications

Antigen CD116
Clone 4H1
Host Species Mouse
Gene Alias GM-CSF Receptor alpha, CSF2RA
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The 4H1 monoclonal antibody reacts with the human CD116 molecule, the α subunit of GM-CSF receptor. The α subunit associates with the common β chain (CD131) to form the high affinity receptor for GM-CSF. The GM-CSFR α chain is expressed by granulocytes, monocytes, endothelial cells, fibroblasts and some tumor cells.