missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD115 (c-fms), PE-eFluor 610, clone: AFS98, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS61115282

 View more versions of this product

Catalog No. 501124777



Description

Description

The protein encoded by this gene is the receptor for colony stimulating factor 1, a cytokine which controls the production, differentiation, and function of macrophages. This receptor mediates most if not all of the biological effects of this cytokine. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. The encoded protein is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in this gene have been associated with a predisposition to myeloid malignancy. The first intron of this gene contains a transcriptionally inactive ribosomal protein L7 processed pseudogene oriented in the opposite direction.

Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. The protein encoded by this gene is the receptor for colony stimulating factor 1, a cytokine which controls the production, differentiation, and function of macrophages. This receptor mediates most if not all of the biological effects of this cytokine. Ligand binding activates the receptor kinase through a process of oligomerization and transphosphorylation. The encoded protein is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. Mutations in this gene have been associated with a predisposition to myeloid maligcy. The first intron of this gene contains a transcriptionally inactive ribosomal protein L7 processed pseudogene oriented in the opposite direction.
Specifications

Specifications

CD115 (c-fms)
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P09581
C-FMS, CD115, CSFR, FIM2, FMS
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
AFS98
PE-eFluor® 610
Csf1r
Liquid
Csf1r
IgG2a, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
12978
Documents

For Research Use Only.