Please login to your online account to display your discounted pricing

CD115 (c-fms) Rat anti-Mouse, APC-eFluor 780, Clone: AFS98, eBioscience™

Rat Monoclonal Antibody

$191.40 - $442.25

Specifications

Antigen CD115 (c-fms)
Clone AFS98
Host Species Rat
Gene Alias FMS, Colony-Stimulating Factor 1 Receptor, M-CSF Receptor
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124041

 
life technologies
LS47115280
25μg Each for $191.40

501129227

 
life technologies
LS47115282
100μg Each for $442.25
Description & Specifications

Specifications

Antigen CD115 (c-fms)
Clone AFS98
Host Species Rat
Gene Alias FMS, Colony-Stimulating Factor 1 Receptor, M-CSF Receptor
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC-eFluor 780
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The AFS98 monoclonal antibody reacts with the mouse CD115 molecule, a receptor for macrophage colony stimulating factor (M-CSF) or colony stimulating factor-1 (CSF-1). CD115 is expressed by monocyte, macrophage, osteoclast, and some epithelial cells. It is a 150kDa c-fms gene product and belongs to immunoglobulin family. CSF-1 signaling through CSF-1R regulates the proliferation and differentiation of cells in the monocytic lineage.