Please login to your online account to display your discounted pricing

CD107b (LAMP-2) Rat anti-Mouse, FITC, Clone: eBioABL-93 (ABL-93), eBioscience™

Rat Monoclonal Antibody

$129.05 - $419.05

Specifications

Antigen CD107b (LAMP-2)
Clone eBioABL-93 (ABL-93)
Host Species Rat
Gene Alias LAMP2, lysosomal-associated membrane protein 2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509732

 
affymetrix
11107281
50μg Each for $129.05

509733

 
affymetrix
11107282
100μg Each for $245.05

509734

 
affymetrix
11107285
500μg Each for $419.05
Description & Specifications

Specifications

Antigen CD107b (LAMP-2)
Clone eBioABL-93 (ABL-93)
Host Species Rat
Gene Alias LAMP2, lysosomal-associated membrane protein 2
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The eBioABL-93 monoclonal antibody reacts with mouse CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated protein of approximately 120kDa that is expressed intracellularly in lysosomal membranes. It is also transiently expressed on the surface of cytolytic T cells during degranulation, but to a lesser extent than CD107a. It has been reported that CD107b may be identical to the mouse Mac-3 antigen.