missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD107b (LAMP-2) Rat anti-Mouse, eFluor 660, Clone: eBioABL-93, eBioscience™

Rat Monoclonal Antibody

$143.55 - $317.55

Specifications

Antigen CD107b (LAMP-2)
Clone eBioABL-93
Host Species Rat
Gene Alias LAMP2, lysosomal-associated membrane protein 2
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124295 Life Technologies
LS50107280
25μg Each for $143.55
501124296 Life Technologies
LS50107282
100μg Each for $317.55
Specifications

Description

The eBioABL-93 monoclonal antibody reacts with mouse CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated protein of approximately 120kDa that is expressed intracellularly in lysosomal membranes. It is also transiently expressed on the surface of cytolytic T cells during degranulation, but to a lesser extent than CD107a. It has been reported that CD107b may be identical to the mouse Mac-3 antigen.
Description & Specifications

Specifications

CD107b (LAMP-2)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioABL-93
LAMP2, lysosomal-associated membrane protein 2
Flow Cytometry (Intracellular Staining)
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL