missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD107b (LAMP-2) Rat anti-Mouse, Clone: eBioABL-93 (ABL-93), eBioscience™

Rat Monoclonal Antibody

$158.05 - $332.05

Specifications

Antigen CD107b (LAMP-2)
Clone eBioABL-93 (ABL-93)
Host Species Rat
Gene Alias LAMP2, lysosomal-associated membrane protein 2
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012814 Life Technologies
LS14107282
100μg Each for $158.05
5012815 Life Technologies
LS14107285
500μg Each for $332.05
Specifications

Description

The eBioABL-93 monoclonal antibody reacts with mouse CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated protein of approximately 120kDa that is expressed intracellularly in lysosomal membranes. It is also transiently expressed on the surface of cytolytic T cells during degranulation, but to a lesser extent than CD107a. It has been reported that CD107b may be identical to the mouse Mac-3 antigen.
Description & Specifications

Specifications

CD107b (LAMP-2)
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioABL-93 (ABL-93)
LAMP2, lysosomal-associated membrane protein 2
Flow Cytometry (Intracellular Staining),Immunohistochemistry,Immunoprecipitation
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL