Please login to your online account to display your discounted pricing

CD107b (LAMP-2) Rat anti-Mouse, Alexa Fluor 488, Clone: ABL-93, eBioscience™

Rat Monoclonal Antibody

$143.55 - $317.55

Specifications

Antigen CD107b (LAMP-2)
Clone eBioABL-93 (ABL-93)
Host Species Rat
Gene Alias LAMP2, lysosomal-associated membrane protein 2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124589

 
life technologies
LS53107280
25μg Each for $143.55

5016719

 
life technologies
LS53107282
100μg Each for $317.55
Description & Specifications

Specifications

Antigen CD107b (LAMP-2)
Clone eBioABL-93 (ABL-93)
Host Species Rat
Gene Alias LAMP2, lysosomal-associated membrane protein 2
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate Alexa Fluor 488
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The eBioABL-93 monoclonal antibody reacts with mouse CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated protein of approximately 120kDa that is expressed intracellularly in lysosomal membranes. It is also transiently expressed on the surface of cytolytic T cells during degranulation, but to a lesser extent than CD107a. It has been reported that CD107b may be identical to the mouse Mac-3 antigen.