Please login to your online account to display your discounted pricing

CD107b (LAMP-2) Mouse anti-Human, FITC, Clone: eBioH4B4 (H4B4), eBioscience™

Mouse Monoclonal Antibody

$143.55 - $288.55

Specifications

Antigen CD107b (LAMP-2)
Clone eBioH4B4 (H4B4)
Host Species Mouse
Gene Alias LAMP2, lysosomal-associated membrane protein 2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509735

 
life technologies
LS11107842
100 tests Each for $288.55

509736

 
life technologies
LS11107841
25 tests Each for $143.55
Description & Specifications

Specifications

Antigen CD107b (LAMP-2)
Clone eBioH4B4 (H4B4)
Host Species Mouse
Gene Alias LAMP2, lysosomal-associated membrane protein 2
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The eBioH4B4 monoclonal antibody reacts with human CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated, type I transmembrane protein of approximately 105 kDa. It is expressed intracellularly in lysosomal/endosomal membranes in nearly all cells. It is also expressed on the surface of degranulating T cells (to a lesser extent than CD107a) and activated platelets as well as some cancer cells. In humans, mutations in CD107b results in a lysosomal glycogen storage disorder, known as Danon disease.