missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD107b (LAMP-2) Mouse anti-Human, eFluor 660, Clone: eBioH4B4 (H4B4), eBioscience™

Mouse Monoclonal Antibody

$134.64 - $340.56

Specifications

Antigen CD107b (LAMP-2)
Clone eBioH4B4 (H4B4)
Host Species Murine
Gene Alias LAMP2, lysosomal-associated membrane protein 2
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124297 Life Technologies
LS50107841
25 Tests Each for $134.64
501124298 Life Technologies
LS50107842
100 Tests Each for $340.56
Specifications

Description

The eBioH4B4 monoclonal antibody reacts with human CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated, type I transmembrane protein of approximately 105 kDa. It is expressed intracellularly in lysosomal/endosomal membranes in nearly all cells. It is also expressed on the surface of degranulating T cells (to a lesser extent than CD107a) and activated platelets as well as some cancer cells. In humans, mutations in CD107b results in a lysosomal glycogen storage disorder, known as Danon disease.
Description & Specifications

Specifications

CD107b (LAMP-2)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioH4B4 (H4B4)
LAMP2, lysosomal-associated membrane protein 2
Flow Cytometry (Intracellular Staining)
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test