missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD107b (LAMP-2) Mouse anti-Human, Clone: eBioH4B4 (H4B4), eBioscience™

Mouse Monoclonal Antibody

$85.55 - $158.05

Specifications

Antigen CD107b (LAMP-2)
Clone eBioH4B4 (H4B4)
Host Species Murine
Gene Alias LAMP2, lysosomal-associated membrane protein 2
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012816 Life Technologies
LS14107880
25μg Each for $85.55
5012817 Life Technologies
LS14107882
100μg Each for $158.05
Specifications

Description

The eBioH4B4 monoclonal antibody reacts with human CD107b, also known as lysosomal-associated membrane protein-2 (LAMP-2). CD107b is a highly glycosylated, type I transmembrane protein of approximately 105 kDa. It is expressed intracellularly in lysosomal/endosomal membranes in nearly all cells. It is also expressed on the surface of degranulating T cells (to a lesser extent than CD107a) and activated platelets as well as some cancer cells. In humans, mutations in CD107b results in a lysosomal glycogen storage disorder, known as Danon disease.
Description & Specifications

Specifications

CD107b (LAMP-2)
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioH4B4 (H4B4)
LAMP2, lysosomal-associated membrane protein 2
Flow Cytometry (Intracellular Staining),Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL